CAS RN 179897-94-0
IUPAC Name:
(5-fluoro-2-methoxyphenyl)boronic acid
Synonyms:
5-fluoro-2-methoxyphenylboronic acid
Molecular Weight (g/mol):
169.95
Molecular Formula:
C7H8BFO3
InChi Key:
CCQKIRUMTHHPSX-UHFFFAOYSA-N
SMILES:
COC1=CC=C(F)C=C1B(O)O
1
–
1
of
1
results
1
5-Fluoro-2-methoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
CAS: 179897-94-0 Molecular Formula: C7H8BFO3 MDL Number: MFCD01863526 InChI Key: CCQKIRUMTHHPSX-UHFFFAOYSA-N PubChem CID: 2782673 IUPAC Name: (5-fluoro-2-methoxyphenyl)boronic acid