accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 180-84-7

IUPAC Name: 1,7-dioxaspiro[5.5]undecane
Synonyms: 1,7-dioxaspiro[5.5]undecane
Molecular Weight (g/mol): 156.23
Molecular Formula: C9H16O2
InChi Key: GBBVHDGKDQAEOT-UHFFFAOYSA-N
SMILES: C1CCC2(CCCCO2)OC1
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)