accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOONNOHO

CAS RN 1802-30-8

IUPAC Name: [2,2'-bipyridine]-5,5'-dicarboxylic acid
Synonyms: [2,2'-bipyridine]-5,5'-dicarboxylic acid
Molecular Weight (g/mol): 244.21
Molecular Formula: C12H8N2O4
InChi Key: KVQMUHHSWICEIH-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(N=C1)C1=CC=C(C=N1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results