accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOOH3CNH2HCl

CAS RN 180258-45-1

IUPAC Name: methyl 4-amino-1-methyl-1H-pyrrole-2-carboxylate hydrochloride
Synonyms: methyl 4-amino-1-methylpyrrole-2-carboxylate hydrochloride
Molecular Weight (g/mol): 190.63
Molecular Formula: C7H11ClN2O2
InChi Key: HEOKCJUUKIPIMM-UHFFFAOYSA-N
SMILES: Cl.COC(=O)C1=CC(N)=CN1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results