accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaH3CSiCH3CH3O

CAS RN 18027-10-6

IUPAC Name: sodium trimethylsilanolate
Synonyms: sodium trimethylsilanolate
Molecular Weight (g/mol): 112.18
Molecular Formula: C3H9NaOSi
InChi Key: HSNUIYJWTSJUMS-UHFFFAOYSA-N
SMILES: [Na+].C[Si](C)(C)[O-]
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)