accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3COSiOSiH3CCH3OSiCH3H3C

CAS RN 18027-45-7

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 327.65
Molecular Formula: C12H23O3Si4
InChi Key: YKSADNUOSVJOAS-UHFFFAOYSA-N
SMILES: C[Si](C)O[Si](O[Si](C)C)(O[Si](C)C)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results