accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 180403-13-8

IUPAC Name: 2-(piperidin-1-yl)-1,3-thiazole-5-carboxylic acid
Synonyms: 2-(piperidin-1-yl)-1,3-thiazole-5-carboxylic acid
Molecular Weight (g/mol): 212.27
Molecular Formula: C9H12N2O2S
InChi Key: JIYHGZWZWANYAV-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=C(S1)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results