accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSiCH3ClCl

CAS RN 18051-88-2

IUPAC Name: dichloro(decyl)methylsilane
Synonyms: dichloro(decyl)methylsilane
Molecular Weight (g/mol): 255.3
Molecular Formula: C11H24Cl2Si
InChi Key: UFVFNJHZBMHRCO-UHFFFAOYSA-N
SMILES: CCCCCCCCCC[Si](C)(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results