accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNOSBr

CAS RN 180530-13-6

IUPAC Name: 3-(5-bromothiophen-2-yl)-5-methyl-1,2,4-oxadiazole
Synonyms: 3-(5-bromothiophen-2-yl)-5-methyl-1,2,4-oxadiazole
Molecular Weight (g/mol): 245.09
Molecular Formula: C7H5BrN2OS
InChi Key: ZNWQLJYQXULFQB-UHFFFAOYSA-N
SMILES: CC1=NC(=NO1)C1=CC=C(Br)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results