accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOBr

CAS RN 180597-83-5

IUPAC Name: 4-(bromomethyl)-5-methyl-3-phenyl-1,2-oxazole
Synonyms: 4-(bromomethyl)-5-methyl-3-phenyl-1,2-oxazole
Molecular Weight (g/mol): 252.11
Molecular Formula: C11H10BrNO
InChi Key: UICMWXWMCOJBIQ-UHFFFAOYSA-N
SMILES: CC1=C(CBr)C(=NO1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results