accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1806-23-1

IUPAC Name: (2-chlorophenyl)(4-fluorophenyl)methanone
Synonyms: (2-chlorophenyl)(4-fluorophenyl)methanone
Molecular Weight (g/mol): 234.65
Molecular Formula: C13H8ClFO
InChi Key: DODIKYQYCCFWRZ-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C(=O)C1=CC=CC=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results