accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1806-26-4

IUPAC Name: 4-octylphenol
Synonyms: 4-octylphenol
Molecular Weight (g/mol): 206.33
Molecular Formula: C14H22O
InChi Key: NTDQQZYCCIDJRK-UHFFFAOYSA-N
SMILES: CCCCCCCCC1=CC=C(O)C=C1
Boiling Point 
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)