accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1806-29-7

IUPAC Name: [1,1'-biphenyl]-2,2'-diol
Synonyms: 2,2'-biphenol o-dihydroxydiphenyl o,o'-diphenol 2,2'-dihydroxydiphenyl o,o'-biphenol O,O'-biphenol
Molecular Weight (g/mol): 186.21
Molecular Formula: C12H10O2
InChi Key: IMHDGJOMLMDPJN-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1C1=CC=CC=C1O
Boiling Point 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (4)
  • (2)
  • (3)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (3)
  • (5)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (4)
  • (2)
  • (3)
  • (3)
  • (2)