accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1806-34-4

IUPAC Name: 5-phenyl-2-[4-(5-phenyl-1,3-oxazol-2-yl)phenyl]-1,3-oxazole
Synonyms: popop
Molecular Weight (g/mol): 364.4
Molecular Formula: C24H16N2O2
InChi Key: MASVCBBIUQRUKL-UHFFFAOYSA-N
SMILES: O1C(=CN=C1C1=CC=C(C=C1)C1=NC=C(O1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)