accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2CNCS

CAS RN 18060-79-2

IUPAC Name: 5-isothiocyanatopent-1-ene
Synonyms: 1-pentene, 5-isothiocyanato-
Molecular Weight (g/mol): 127.21
Molecular Formula: C6H9NS
InChi Key: DBISBKDNOKIADM-UHFFFAOYSA-N
SMILES: C=CCCCN=C=S
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results