accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOOCH3

CAS RN 18065-13-9

IUPAC Name: 1-(4-methoxybenzoyl)-4-methylpiperidine
Synonyms: 1-(4-methoxybenzoyl)-4-methylpiperidine
Molecular Weight (g/mol): 233.31
Molecular Formula: C14H19NO2
InChi Key: VUAQWLKBOXJFKX-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)N1CCC(C)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results