accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1807685-51-3

IUPAC Name: 1-[3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethan-1-one
Synonyms: 1-[3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone
Molecular Weight (g/mol): 264.1
Molecular Formula: C14H18BFO3
InChi Key: QRPSPYZFNHVURF-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(F)=C(C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results