CAS RN 1807776-35-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tri(cyclohexa-2,5-dien-1-yl)silane 93.0+%, TCI America™
CAS: 1807776-35-7 Molecular Formula: C18H22Si