accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONHOO

CAS RN 180798-01-0

IUPAC Name: [(benzyloxy)carbonyl]amino acetate
Synonyms: [(benzyloxy)carbonyl]amino acetate
Molecular Weight (g/mol): 209.2
Molecular Formula: C10H11NO4
InChi Key: YEIRDDKOGLZHLP-UHFFFAOYSA-N
SMILES: CC(=O)ONC(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results