CAS RN 18092-54-1
IUPAC Name:
4-methoxy-2-nitrobenzene-1-sulfonyl chloride
Synonyms:
4-methoxy-2-nitrobenzenesulfonyl chloride
Molecular Weight (g/mol):
251.64
Molecular Formula:
C7H6ClNO5S
InChi Key:
OGHAPVZVXLHURO-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C(=C1)[N+]([O-])=O)S(Cl)(=O)=O
1
–
1
of
1
results
1
4-Methoxy-2-nitrobenzenesulfonyl Chloride 93.0+%, TCI America™
CAS: 18092-54-1 Molecular Formula: C7H6ClNO5S Molecular Weight (g/mol): 251.637 MDL Number: MFCD00143333 InChI Key: OGHAPVZVXLHURO-UHFFFAOYSA-N PubChem CID: 87452 IUPAC Name: 4-methoxy-2-nitrobenzenesulfonyl chloride SMILES: COC1=CC(=C(C=C1)S(=O)(=O)Cl)[N+](=O)[O-]