CAS RN 1809298-92-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,3-Difluoro-4-methoxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific™
CAS: 1809298-92-7 Synonym: 2,3-Difluoro-4-methoxyphenylboronic acid pinacol ester