CAS RN 180995-12-4
IUPAC Name:
2,4-dichloropyridine-3-carbonitrile
Synonyms:
2,4-dichloropyridine-3-carbonitrile
Molecular Weight (g/mol):
173
Molecular Formula:
C6H2Cl2N2
InChi Key:
OQYZEFBWEXEOPL-UHFFFAOYSA-N
SMILES:
ClC1=C(C#N)C(Cl)=NC=C1
1
–
1
of
1
results
1
Promotions Available
2,4-Dichloro-3-cyanopyridine, 95%
2,4-Dichloro-3-cyanopyridine, 95%
CAS: 180995-12-4 Molecular Formula: C6H2Cl2N2 Molecular Weight (g/mol): 172.996 MDL Number: MFCD09750302 InChI Key: OQYZEFBWEXEOPL-UHFFFAOYSA-N PubChem CID: 21997828 IUPAC Name: 2,4-dichloropyridine-3-carbonitrile SMILES: C1=CN=C(C(=C1Cl)C#N)Cl