accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1810-13-5

IUPAC Name: 3,3,4,4-tetrafluoropyrrolidine hydrochloride
Synonyms: 3,3,4,4-tetrafluoropyrrolidine hydrochloride
Molecular Weight (g/mol): 179.54
Molecular Formula: C4H6ClF4N
InChi Key: KPTMFLKIYHWQPB-UHFFFAOYSA-N
SMILES: Cl.FC1(F)CNCC1(F)F
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results