accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1810-71-5

IUPAC Name: 6-bromo-2-chloroquinoline
Synonyms: 6-bromo-2-chloroquinoline
Molecular Weight (g/mol): 242.5
Molecular Formula: C9H5BrClN
InChi Key: YXRDWUJAJLDABJ-UHFFFAOYSA-N
SMILES: ClC1=NC2=CC=C(Br)C=C2C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)