accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 181259-35-8

IUPAC Name: tetrakis(4-methylphenyl)boranuide; tetraphenylphosphanium
Synonyms: tetrakis(4-methylphenyl)boranuide; tetraphenylphosphonium
Molecular Weight (g/mol): 714.74
Molecular Formula: C52H48BP
InChi Key: FAYPBNFHFCEUQZ-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1.CC1=CC=C(C=C1)[B-](C1=CC=C(C)C=C1)(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results