accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOOCCCH3CH3CCH3H3CCH3H3CHHHEu

CAS RN 181266-82-0

IUPAC Name: europium(3+) tris(2,4-dioxopentan-3-ide)
Synonyms: europium acetylacetonate
Molecular Weight (g/mol): 449.29
Molecular Formula: C15H21EuO6
InChi Key: YMTKZYSHNNXTIJ-UHFFFAOYSA-N
SMILES: [Eu+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)