accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3O

CAS RN 18127-01-0

IUPAC Name: 3-(4-tert-butylphenyl)propanal
Synonyms: bourgeonal
Molecular Weight (g/mol): 190.29
Molecular Formula: C13H18O
InChi Key: FZJUFJKVIYFBSY-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(CCC=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results