accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3NCH3

CAS RN 18138-04-0

IUPAC Name: 2,3-diethyl-5-methylpyrazine
Synonyms: 2,3-diethyl-5-methylpyrazine
Molecular Weight (g/mol): 150.23
Molecular Formula: C9H14N2
InChi Key: PSINWXIDJYEXLO-UHFFFAOYSA-N
SMILES: CCC1=NC=C(C)N=C1CC
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results