accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1817-49-8

IUPAC Name: 1,3-diphenylprop-2-yn-1-ol
Synonyms: 1,3-diphenylprop-2-yn-1-ol
Molecular Weight (g/mol): 208.26
Molecular Formula: C15H12O
InChi Key: DZZWMODRWHHWFR-UHFFFAOYNA-N
SMILES: OC(C#CC1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results