accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO

CAS RN 1817-57-8

IUPAC Name: 4-phenylbut-3-yn-2-one
Synonyms: 4-phenyl-3-butyn-2-one
Molecular Weight (g/mol): 144.17
Molecular Formula: C10H8O
InChi Key: UPEUQDJSUFHFQP-UHFFFAOYSA-N
SMILES: CC(=O)C#CC1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)