accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONONOO

CAS RN 1817-74-9

IUPAC Name: 1-nitro-4-[(4-nitrophenyl)methyl]benzene
Synonyms: 1-nitro-4-[(4-nitrophenyl)methyl]benzene
Molecular Weight (g/mol): 258.23
Molecular Formula: C13H10N2O4
InChi Key: GLBZQZXDUTUCGK-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(CC2=CC=C(C=C2)[N+]([O-])=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results