CAS RN 181934-29-2
IUPAC Name:
1-[2-(trimethylsilyl)ethynyl]-3H-1λ³,2-benziodaoxol-3-one
Synonyms:
1-[2-(trimethylsilyl)ethynyl]-1λ³,2-benziodaoxol-3-one
Molecular Weight (g/mol):
344.22
Molecular Formula:
C12H13IO2Si
InChi Key:
KUCCHAADRXRBFZ-UHFFFAOYSA-N
SMILES:
C[Si](C)(C)C#C[I]1OC(=O)C2=CC=CC=C12
1
–
1
of
1
results
1
1-[(Trimethylsilyl)ethynyl]-1,2-benziodoxol-3(1H)-one 98.0+%, TCI America™
CAS: 181934-29-2 Molecular Formula: C12H13IO2Si Molecular Weight (g/mol): 344.223 InChI Key: KUCCHAADRXRBFZ-UHFFFAOYSA-N Synonym: TMS-EBX PubChem CID: 10641155 IUPAC Name: 1-(2-trimethylsilylethynyl)-1$l^{3},2-benziodoxol-3-one SMILES: C[Si](C)(C)C#CI1C2=CC=CC=C2C(=O)O1