accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3SiIOOH3CH3CCH3CH3

CAS RN 181934-30-5

IUPAC Name: 1-{2-[tris(propan-2-yl)silyl]ethynyl}-3H-1λ³,2-benziodaoxol-3-one
Synonyms: 1-[2-(triisopropylsilyl)ethynyl]-1λ³,2-benziodaoxol-3-one
Molecular Weight (g/mol): 428.39
Molecular Formula: C18H25IO2Si
InChi Key: NTHGHMCOPNSZIR-UHFFFAOYSA-N
SMILES: CC(C)[Si](C#C[I]1OC(=O)C2=CC=CC=C12)(C(C)C)C(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results