accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO(S)OPOOONH3CCH3H3COOCH3

CAS RN 18194-24-6

IUPAC Name: (2-{[(2S)-2,3-bis(tetradecanoyloxy)propyl phosphonato]oxy}ethyl)trimethylazanium
Synonyms: (2-{[(2S)-2,3-bis(tetradecanoyloxy)propyl phosphonato]oxy}ethyl)trimethylazanium
Molecular Weight (g/mol): 677.95
Molecular Formula: C36H72NO8P
InChi Key: CITHEXJVPOWHKC-UMSFTDKQSA-N
SMILES: CCCCCCCCCCCCCC(=O)OC[C@@H](COP([O-])(=O)OCC[N+](C)(C)C)OC(=O)CCCCCCCCCCCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results