accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18197-26-7

IUPAC Name: sodium diformylazanide
Synonyms: sodium diformylazanide
Molecular Weight (g/mol): 95.03
Molecular Formula: C2H2NNaO2
InChi Key: QJXDSDLNUKLDBP-UHFFFAOYSA-M
SMILES: [Na+].O=C[N-]C=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)