accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClCH3H3CCH3NH2HN

CAS RN 18202-73-8

IUPAC Name: hydrogen 2,2-dimethylpropanimidamide chloride
Synonyms: hydrogen 2,2-dimethylpropanimidamide chloride
Molecular Weight (g/mol): 136.62
Molecular Formula: C5H13ClN2
InChi Key: ARDGQYVTLGUJII-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC(C)(C)C(N)=N
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)