accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiClClH3C

CAS RN 18204-93-8

IUPAC Name: dichlorodihexylsilane
Synonyms: dichlorodihexylsilane
Molecular Weight (g/mol): 269.33
Molecular Formula: C12H26Cl2Si
InChi Key: NRAYZPGATNMOSB-UHFFFAOYSA-N
SMILES: CCCCCC[Si](Cl)(Cl)CCCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results