accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOOOOOCH3OONOOOOCH3OOH3C

CAS RN 1820575-44-7

IUPAC Name: [3,4-bis(acetyloxy)-6-{[3,5-bis(benzyloxy)-2-[(benzyloxy)methyl]-6-(4-methoxyphenoxy)oxan-4-yl]oxy}-5-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)oxan-2-yl]methyl acetate
Synonyms: [3,4-bis(acetyloxy)-6-{[3,5-bis(benzyloxy)-2-[(benzyloxy)methyl]-6-(4-methoxyphenoxy)oxan-4-yl]oxy}-5-(1,3-dioxoisoindol-2-yl)oxan-2-yl]methyl acetate
Molecular Weight (g/mol): 974.03
Molecular Formula: C54H55NO16
InChi Key: QHFVNDYHDLXLJC-UHFFFAOYNA-N
SMILES: COC1=CC=C(OC2OC(COCC3=CC=CC=C3)C(OCC3=CC=CC=C3)C(OC3OC(COC(C)=O)C(OC(C)=O)C(OC(C)=O)C3N3C(=O)C4=CC=CC=C4C3=O)C2OCC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results