accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1820580-75-3

H3COOOOOH3COHOOH3CH3C

CAS RN 1820580-75-3

IUPAC Name: [7-hydroxy-6-(4-methoxyphenoxy)-2,2-dimethyl-hexahydro-[1,3]dioxolo[4,5-c]pyran-4-yl]methyl 4-methylbenzoate
Synonyms: [7-hydroxy-6-(4-methoxyphenoxy)-2,2-dimethyl-tetrahydro-3aH-[1,3]dioxolo[4,5-c]pyran-4-yl]methyl 4-methylbenzoate
Molecular Weight (g/mol): 444.48
Molecular Formula: C24H28O8
InChi Key: WHYAFYRMODYSLP-UHFFFAOYNA-N
SMILES: COC1=CC=C(OC2OC(COC(=O)C3=CC=C(C)C=C3)C3OC(C)(C)OC3C2O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)