accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1820954-21-9

IUPAC Name: (4R,5S,6S)-6-[(1R)-1-hydroxyethyl]-4-methyl-7-oxo-3-{[(3S,5S)-5-[(sulfamoylamino)methyl]pyrrolidin-3-yl]sulfanyl}-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid
Synonyms: doripenem
Molecular Weight (g/mol): 420.5
Molecular Formula: C15H24N4O6S2
InChi Key: AVAACINZEOAHHE-SEIFCSKWNA-N
SMILES: [H][C@]12[C@@H](C)C(S[C@@H]3CN[C@H](CNS(N)(=O)=O)C3)=C(N1C(=O)[C@]2([H])[C@@H](C)O)C(O)=O
Quantity 
  • (1)
  • (1)
Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results