accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1821-02-9

IUPAC Name: 2-oxopentanoic acid
Synonyms: α-oxopentanoic acid
Molecular Weight (g/mol): 116.12
Molecular Formula: C5H8O3
InChi Key: KDVFRMMRZOCFLS-UHFFFAOYSA-N
SMILES: CCCC(=O)C(O)=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)