accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOHNSNNH2H2NHCl

CAS RN 182153-75-9

IUPAC Name: N''-[3-(5-oxo-1-phenyl-2-sulfanylideneimidazolidin-4-yl)propyl]guanidine hydrochloride
Synonyms: N''-[3-(5-oxo-1-phenyl-2-sulfanylideneimidazolidin-4-yl)propyl]guanidine hydrochloride
Molecular Weight (g/mol): 327.83
Molecular Formula: C13H18ClN5OS
InChi Key: IXKPSQGERWCHMF-UHFFFAOYNA-N
SMILES: Cl.NC(N)=NCCCC1NC(=S)N(C1=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results