accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3SiOOOCH3OSiH3CCH3H3C

CAS RN 182250-68-6

IUPAC Name: methyl 3,5-bis[(tert-butyldiphenylsilyl)oxy]benzoate
Synonyms: methyl 3,5-bis[(tert-butyldiphenylsilyl)oxy]benzoate
Molecular Weight (g/mol): 644.96
Molecular Formula: C40H44O4Si2
InChi Key: DGYIOKNASZQETF-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(O[Si](C2=CC=CC=C2)(C2=CC=CC=C2)C(C)(C)C)=CC(O[Si](C2=CC=CC=C2)(C2=CC=CC=C2)C(C)(C)C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results