accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOOO

CAS RN 1823-59-2

IUPAC Name: 5-[(1,3-dioxo-1,3-dihydro-2-benzofuran-5-yl)oxy]-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms: 5-[(1,3-dioxo-2-benzofuran-5-yl)oxy]-2-benzofuran-1,3-dione
Molecular Weight (g/mol): 310.22
Molecular Formula: C16H6O7
InChi Key: QQGYZOYWNCKGEK-UHFFFAOYSA-N
SMILES: O=C1OC(=O)C2=CC(OC3=CC=C4C(=O)OC(=O)C4=C3)=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)