accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OClOCH3F

CAS RN 1823049-90-6

IUPAC Name: 5-chloro-3-fluoro-4-methoxy-2-methylbenzaldehyde
Synonyms: 5-chloro-3-fluoro-4-methoxy-2-methylbenzaldehyde
Molecular Weight (g/mol): 202.61
Molecular Formula: C9H8ClFO2
InChi Key: VILWOSMVUDTFHT-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C=C(C=O)C(C)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results