accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 182344-13-4

BOHClHOOH

CAS RN 182344-13-4

IUPAC Name: (3-chloro-4-hydroxyphenyl)boronic acid
Synonyms: 3-chloro-4-hydroxyphenylboronic acid
Molecular Weight (g/mol): 172.37
Molecular Formula: C6H6BClO3
InChi Key: WWQIKFZZILXJHG-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(Cl)=C(O)C=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results