accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOHFHOOH

CAS RN 182344-14-5

IUPAC Name: (3-fluoro-4-hydroxyphenyl)boronic acid
Synonyms: 3-fluoro-4-hydroxyphenylboronic acid
Molecular Weight (g/mol): 155.92
Molecular Formula: C6H6BFO3
InChi Key: OYNDLOJPYURCJG-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(F)=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results