accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOBOHClFFF

CAS RN 182344-18-9

IUPAC Name: [2-chloro-5-(trifluoromethyl)phenyl]boronic acid
Synonyms: 2-chloro-5-(trifluoromethyl)phenylboronic acid
Molecular Weight (g/mol): 224.37
Molecular Formula: C7H5BClF3O2
InChi Key: YVMXEHZEYONARR-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CC=C1Cl)C(F)(F)F
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results