accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H2NN

CAS RN 1824-81-3

IUPAC Name: 6-methylpyridin-2-amine
Synonyms: 2-pyridinamine, 6-methyl-
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: QUXLCYFNVNNRBE-UHFFFAOYSA-N
SMILES: CC1=CC=CC(N)=N1
Boiling Point 
  • (6)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (5)
  • (2)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (3)
  • (6)
  • (6)
  • (1)
  • (3)
  • (3)
  • (5)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (3)
  • (1)