CAS RN 18242-39-2
IUPAC Name:
1-bromo-3,5-dinitrobenzene
Synonyms:
1-bromo-3,5-dinitrobenzene
Molecular Weight (g/mol):
247
Molecular Formula:
C6H3BrN2O4
InChi Key:
OLDMYNWXIGPOCI-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC(=CC(Br)=C1)[N+]([O-])=O
1
–
1
of
1
results
1
Promotions Available
1-Bromo-3,5-dinitrobenzene, 98%
1-Bromo-3,5-dinitrobenzene, 98%
CAS: 18242-39-2 Molecular Formula: C6H3BrN2O4 Molecular Weight (g/mol): 247.004 MDL Number: MFCD00156596 InChI Key: OLDMYNWXIGPOCI-UHFFFAOYSA-N PubChem CID: 44534 IUPAC Name: 1-bromo-3,5-dinitrobenzene SMILES: C1=C(C=C(C=C1[N+](=O)[O-])Br)[N+](=O)[O-]